CH6371385563 Chem Impex International, Inc.

Boc-3-amino-3-methyl-butyric acid

  • MDL number MFCD09031500
  • Cas Number : 129765-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-101
Purity 96
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-3-amino-3-methyl-butyric-acid/13327?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-3-amino-3-methyl-butyric-acid/13327?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart