CH6371385600 Chem Impex International, Inc.

Sodium bismuthate

  • MDL number MFCD00003474
  • Cas Number : 12232-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Bismuth sodium trioxide
Purity 80.0
Storage temp. Store at RT
Physical Form Yellow to brown powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/sodium-bismuthate/13364?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/sodium-bismuthate/13364?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart