CH6371385655 Chem Impex International, Inc.

Mercury(I) chloride

  • MDL number MFCD00011043
  • Cas Number : 10112-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 400-500 �C
Purity 99.5
Storage temp. Store at RT
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/mercury-i-chloride/13420?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/mercury-i-chloride/13420?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart