CH6371385766 Chem Impex International, Inc.

2-(4-Fluorophenoxy)propylamine

  • MDL number MFCD02684117
  • Cas Number : 886763-30-0
  • Molecular Weight : 169.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Physical Form Liquid
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/2-4-fluorophenoxy-propylamine/14073?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-4-fluorophenoxy-propylamine/14073?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart