CH6371385767 Chem Impex International, Inc.

2-(4-Trifluoromethyl-phenoxy)propylamine

  • MDL number MFCD02684119
  • Cas Number : 886763-49-1
  • Molecular Weight : 219.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 94
Storage temp. Store at 0-8
Physical Form Liquid
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/2-4-trifluoromethyl-phenoxy-propylamine/14074?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-4-trifluoromethyl-phenoxy-propylamine/14074?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart