CH6371385852 Chem Impex International, Inc.

3-[4-(Trifluoromethyl)Phenyl]Morpholine Hydrochloride

  • MDL number MFCD03840092
  • Cas Number : 1185140-30-0
  • Molecular Weight : 267.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Physical Form Yellow powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/3-4-trifluoromethyl-phenyl-morpholine-hydrochloride/14161?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-4-trifluoromethyl-phenyl-morpholine-hydrochloride/14161?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart