CH6371385896 Chem Impex International, Inc.

4-[4-(Trifluoromethyl)Benzyl]Piperidine Hydrochloride

  • MDL number MFCD03840142
  • Cas Number : 193357-81-2
  • Molecular Weight : 279.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/4-4-trifluoromethyl-benzyl-piperidine-hydrochloride/14205?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-4-trifluoromethyl-benzyl-piperidine-hydrochloride/14205?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart