CH6371385986 Chem Impex International, Inc.

(4-Trifluoromethyl-pyridin-2-yl)methylamine hydrochloride

  • MDL number MFCD04117763
  • Cas Number : 1005515-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.99
Storage temp. Store at 0-8
Physical Form Light yellow liquid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-trifluoromethyl-pyridin-2-yl-methylamine-hydrochloride/14295?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-trifluoromethyl-pyridin-2-yl-methylamine-hydrochloride/14295?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart