CH6371386044 Chem Impex International, Inc.

2-Phenoxybenzonitrile

  • MDL number MFCD08271960
  • Cas Number : 6476-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form Colorless liquid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Boiling point 145-150 � 6mm
Product Link http://www.chemimpex.com/product/productinfo/2-phenoxybenzonitrile/14353?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-phenoxybenzonitrile/14353?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart