CH6371386071 Chem Impex International, Inc.

5-(4-Methoxyphenyl)-3,4-dihydro-2H-pyrrole

  • MDL number MFCD09749694
  • Cas Number : 22217-80-7
  • Molecular Weight : 175.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/5-4-methoxyphenyl-3-4-dihydro-2-i-h-i-pyrrole/14381?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-4-methoxyphenyl-3-4-dihydro-2-i-h-i-pyrrole/14381?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart