CH6371386080 Chem Impex International, Inc.

6-Bromobenzo[d]isoxazol-3-ylamine

  • MDL number MFCD09788587
  • Cas Number : 177995-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Bromobenzo[d]isoxazol-3-amine
Purity 98
Storage temp. Store at 0-8
Physical Form Off-white to yellow solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/6-bromobenzo-i-delta-i-isoxazol-3-ylamine/14390?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/6-bromobenzo-i-delta-i-isoxazol-3-ylamine/14390?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart