CH6371386117 Chem Impex International, Inc.

Ethyl 2-(Aminomethyl)Isonicotinate Hydrochloride

  • MDL number MFCD09788627
  • Cas Number : 1189983-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form Off-white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ethyl-2-aminomethyl-isonicotinate-hydrochloride/14428?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ethyl-2-aminomethyl-isonicotinate-hydrochloride/14428?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart