CH6371386164 Chem Impex International, Inc.

(S)-2-(((9H-Fluoren-9-yl)methoxy)carbonylamino)-3-(1,3-dioxoisoindolin-2-yl)propanoic acid

  • MDL number MFCD09910433
  • Cas Number : 1212405-65-6
  • Molecular Weight : 456.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0 - 8
Physical Form White solid
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-2-9-i-h-i-fluoren-9-yl-methoxy-carbonylamino-3-1-3-dioxoisoindolin-2-yl-propanoic-acid/14475?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-2-9-i-h-i-fluoren-9-yl-methoxy-carbonylamino-3-1-3-dioxoisoindolin-2-yl-propanoic-acid/14475?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart