CH6371386167 Chem Impex International, Inc.

3-Tert-Butyl-1-(2,2,2-Trifluoroethyl)-1H-Pyrazol-5-Amine

  • MDL number MFCD09910436
  • Cas Number : 1048389-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-tert-butyl-1-2-2-2-trifluoroethyl-1h-pyrazol-5-amine/14478?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-tert-butyl-1-2-2-2-trifluoroethyl-1h-pyrazol-5-amine/14478?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart