CH6371386354 Chem Impex International, Inc.

N,N-Bis(2-chloroethyl)-p-toluenesulfonamide

  • MDL number MFCD00018944
  • Cas Number : 42137-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45-50
Synonym N,N-Bis(2-chloroethyl)-4-methylbenzenesulfonamide
Purity 99
Storage temp. Store at 0-8
Physical Form Off-white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-i-n-i-bis-2-chloroethyl-i-pi-i-toluenesulfonamide/14676?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-i-n-i-bis-2-chloroethyl-i-pi-i-toluenesulfonamide/14676?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart