CH6371386364 Chem Impex International, Inc.

5-Nitro-6-methyluracil

  • MDL number MFCD00047362
  • Cas Number : 16632-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,4-Dihydroxy-5-nitro-6-methylpyrimidine
Storage temp. Store at 0-8
Physical Form Lighy yellow to yellow solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-nitro-6-methyluracil/14686?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-nitro-6-methyluracil/14686?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart