CH6371386411 Chem Impex International, Inc.

[(3S)-(3-1,2,3,4-Tetrahydroisoquinolyl)]-N-(Tert-Butyl)Carboxamide

  • MDL number MFCD01313227
  • Cas Number : 149182-72-9
  • Molecular Weight : 232.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93-100� C
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/3s-3-1-2-3-4-tetrahydroisoquinolyl-n-tert-butyl-carboxamide/14733?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3s-3-1-2-3-4-tetrahydroisoquinolyl-n-tert-butyl-carboxamide/14733?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart