CH6371386812 Chem Impex International, Inc.

Boc-Asp-OMe

  • MDL number MFCD03094777
  • Cas Number : 98045-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86-91 �C
Synonym Boc-L-aspartic acid alpha-methyl ester
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-asp-ome/15168?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-asp-ome/15168?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart