CH6371387128 Chem Impex International, Inc.

5-(4-Aminophenyl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD00806252
  • Cas Number : 32058-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-(4-Aminophenyl)-1,3,4-oxadiazole-2(3H)-thione
Purity 97
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-4-aminophenyl-1-3-4-oxadiazole-2-thiol/15589?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-4-aminophenyl-1-3-4-oxadiazole-2-thiol/15589?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart