CH6371387273 Chem Impex International, Inc.

5-(4-Nitrophenyl)isoxazole-3-carboxylic acid

  • MDL number MFCD06199331
  • Cas Number : 33282-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214-224 �C
Purity 98
Storage temp. Store at 0-8
Physical Form White to brown solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-4-nitrophenyl-isoxazole-3-carboxylic-acid/16036?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-4-nitrophenyl-isoxazole-3-carboxylic-acid/16036?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart