CH6371387321 Chem Impex International, Inc.

4-(4-Chlorophenyl)-2-pyrimidinethiol

  • MDL number MFCD00174262
  • Cas Number : 175203-08-4
  • Molecular Weight : 222.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 211-219
Purity 98
Storage temp. Store at 0-8
Physical Form Yellow powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/4-4-chlorophenyl-2-pyrimidinethiol/16103?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-4-chlorophenyl-2-pyrimidinethiol/16103?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart