CH6371387662 Chem Impex International, Inc.

2-(5-Isoxazolyl)phenol

  • MDL number MFCD00218653
  • Cas Number : 61348-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-188
Synonym 5-(2-Hydroxyphenyl)isoxazole
Purity 98
Storage temp. Store at 0-8
Physical Form Pale yellow crystals
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-5-isoxazolyl-phenol/16554?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-5-isoxazolyl-phenol/16554?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart