CH6371387844 Chem Impex International, Inc.

Ethyl 2-(3-fluoro-phenyl)-thiazole-5-carboxylate

  • MDL number MFCD07375319
  • Cas Number : 886369-79-5
  • Molecular Weight : 251.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Physical Form White to off-white solid
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/ethyl-2-3-fluoro-phenyl-thiazole-5-carboxylate/16743?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ethyl-2-3-fluoro-phenyl-thiazole-5-carboxylate/16743?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart