CH6371388294 Chem Impex International, Inc.

5-tert-Butyl-3-methyl-1-phenyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD06653322
  • Cas Number : 956758-73-9
  • Molecular Weight : 258.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Physical Form Beige solid
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/5-i-tert-i-butyl-3-methyl-1-phenyl-1h-pyrazole-4-carboxylic-acid/17216?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-i-tert-i-butyl-3-methyl-1-phenyl-1h-pyrazole-4-carboxylic-acid/17216?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart