CH6371388622 Chem Impex International, Inc.

1,2-Dimethoxy-4-nitrobenzene

  • MDL number MFCD00007238
  • Cas Number : 709-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-102
Synonym 4-Nitroveratrole
Purity 99
Storage temp. Store at 0-8
Physical Form Yellow crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-2-dimethoxy-4-nitrobenzene/17553?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-2-dimethoxy-4-nitrobenzene/17553?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart