CH6371388676 Chem Impex International, Inc.

4,4'-Dimethoxydibenzoyl

  • MDL number MFCD00008405
  • Cas Number : 1226-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129-136
Synonym p-Anisil
Purity 99
Storage temp. Store at 0-8
Physical Form Yellow powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-4-dimethoxydibenzoyl/17608?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-4-dimethoxydibenzoyl/17608?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart