CH6371388832 Chem Impex International, Inc.

2,5-Dimethylphenylacetic acid

  • MDL number MFCD00014369
  • Cas Number : 13612-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123-129
Synonym (2,5-Xylyl)acetic acid
Purity 98
Storage temp. Store at 0-8
Physical Form Light beige crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-5-dimethylphenylacetic-acid/17769?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-5-dimethylphenylacetic-acid/17769?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart