CH6371388867 Chem Impex International, Inc.

4,4'-Bisbenzyl alcohol

  • MDL number MFCD00016713
  • Cas Number : 1667-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189-197
Synonym 4,4'-Biphenyldimethanol
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-4-bisbenzyl-alcohol/17804?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-4-bisbenzyl-alcohol/17804?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart