CH6371388992 Chem Impex International, Inc.

4-Chloro-3-nitrophenyl methyl sulfone

  • MDL number MFCD00039754
  • Cas Number : 97-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form White to light yellow solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-chloro-3-nitrophenyl-methyl-sulfone/17932?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-chloro-3-nitrophenyl-methyl-sulfone/17932?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart