CH6371389133 Chem Impex International, Inc.

Phenyl tribromomethyl sulfone

  • MDL number MFCD00060068
  • Cas Number : 17025-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144-151
Purity 99
Storage temp. Store at 0-8
Physical Form White to yellow powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/phenyl-tribromomethyl-sulfone/18079?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/phenyl-tribromomethyl-sulfone/18079?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart