CH6371389273 Chem Impex International, Inc.

2-Amino-3,5-dichloro-4-methylpyridine

  • MDL number MFCD00129031
  • Cas Number : 31430-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123-127
Synonym 2-Amino-3,5-dichloro-4-picoline
Purity 97
Storage temp. Store at 0-8
Physical Form Off-white to tan colored powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-amino-3-5-dichloro-4-methylpyridine/18232?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-amino-3-5-dichloro-4-methylpyridine/18232?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart