CH6371389376 Chem Impex International, Inc.

L-Ribose

  • MDL number MFCD00167010
  • Cas Number : 24259-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Ribofuranose
Purity 98
Storage temp. Store at 0-8
Physical Form White to almost white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-ribose/18338?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-ribose/18338?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart