CH6371389401 Chem Impex International, Inc.

5-Ethyl-1-methyl-1H-1,2,3,4-tetrazole

  • MDL number MFCD00190184
  • Cas Number : 90329-50-3
  • Molecular Weight : 112.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Physical Form Clear, brown liquid
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/5-ethyl-1-methyl-1-i-h-i-1-2-3-4-tetrazole

Safety Information

MSDS http://www.chemimpex.com/5-ethyl-1-methyl-1-i-h-i-1-2-3-4-tetrazole
Availability details for
Quantity
  • Availability
  • Add to Cart