CH6371389465 Chem Impex International, Inc.

2-Bromo-6-nitropyridine

  • MDL number MFCD00234012
  • Cas Number : 21203-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Bromo-2-nitropyridine
Purity 99
Storage temp. Store at 0-8
Physical Form Light brown powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-bromo-6-nitropyridine/18439?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-bromo-6-nitropyridine/18439?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart