CH6371389502 Chem Impex International, Inc.

2-Iodo-5-hydroxybenzoic acid

  • MDL number MFCD00239284
  • Cas Number : 57772-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 193-196
Purity 98
Storage temp. Store at 0-8
Physical Form Off-white to tan powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-iodo-5-hydroxybenzoic-acid/18477?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-iodo-5-hydroxybenzoic-acid/18477?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart