CH6371389701 Chem Impex International, Inc.

3-Iodopyridine-2-carboxylic acid

  • MDL number MFCD01928487
  • Cas Number : 73841-32-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Iodopyridine-2-carboxylic acid
Purity 97
Storage temp. Store at 0-8
Physical Form Yellowish brown solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-iodopyridine-2-carboxylic-acid/18708?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-iodopyridine-2-carboxylic-acid/18708?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart