CH6371389724 Chem Impex International, Inc.

4-(2-Hydroxyethyl)benzoic acid

  • MDL number MFCD02114379
  • Cas Number : 46112-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122-129 �C
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-2-hydroxyethyl-benzoic-acid/18733?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-2-hydroxyethyl-benzoic-acid/18733?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart