CH6371389892 Chem Impex International, Inc.

3-Methyl-4-nitrobenzyl bromide

  • MDL number MFCD04039920
  • Cas Number : 141281-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44-50
Purity 96
Storage temp. Store at 0-8
Physical Form Off-white to light yellow powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-methyl-4-nitrobenzyl-bromide/18922?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-methyl-4-nitrobenzyl-bromide/18922?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart