CH6371389921 Chem Impex International, Inc.

2-Methyl-6-nitropyridine

  • MDL number MFCD04114203
  • Cas Number : 18368-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106-112
Synonym 6-Methyl-2-nitropyridine
Purity 98
Storage temp. Store at 0-8
Physical Form Pale yellow crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-methyl-6-nitropyridine/18951?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-methyl-6-nitropyridine/18951?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart