CH6371389922 Chem Impex International, Inc.

3-Bromo-5-nitropyridine

  • MDL number MFCD04114216
  • Cas Number : 15862-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106-117
Purity 99
Storage temp. Store at 0-8
Physical Form Yellow to brown solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-bromo-5-nitropyridine/18952?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-bromo-5-nitropyridine/18952?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart