CH6371389937 Chem Impex International, Inc.

2-Amino-3-nitro-5-fluoropyridine

  • MDL number MFCD04972413
  • Cas Number : 212268-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149-157
Synonym 2-Amino-5-fluoro-3-nitropyridine
Purity 99
Storage temp. Store at 0-8
Physical Form Off-white to yellow solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-amino-3-nitro-5-fluoropyridine/18968?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-amino-3-nitro-5-fluoropyridine/18968?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart