CH6371390002 Chem Impex International, Inc.

5-Bromo-2-hydroxy-4-methyl-3-nitropyridine

  • MDL number MFCD06659520
  • Cas Number : 228410-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-2-hydroxy-3-nitro-4-picoline
Purity 98.0
Storage temp. Store at 0-8
Physical Form Slightly brown powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-bromo-2-hydroxy-4-methyl-3-nitropyridine/19036?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-bromo-2-hydroxy-4-methyl-3-nitropyridine/19036?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart