CH6371390040 Chem Impex International, Inc.

(3-Hydroxyphenyl)acetic acid ethyl ester

  • MDL number MFCD07369387
  • Cas Number : 22446-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ethyl 3-hydroxyphenylacetate
Purity 99
Storage temp. Store at 0-8
Physical Form Yellow to pale yellow clear liquid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-hydroxyphenyl-acetic-acid-ethyl-ester/19075?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-hydroxyphenyl-acetic-acid-ethyl-ester/19075?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart