CH6371390042 Chem Impex International, Inc.

(S)-2-(3-((2-Isopropylthiazol-4-yl)methyl)-3-methylureido)-3-methylbutanoic acid

  • MDL number MFCD07369527
  • Cas Number : 154212-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form Off-white to yellow solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-2-3-2-isopropylthiazol-4-yl-methyl-3-methylureido-3-methylbutanoic-acid/19077?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-2-3-2-isopropylthiazol-4-yl-methyl-3-methylureido-3-methylbutanoic-acid/19077?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart