CH6371390047 Chem Impex International, Inc.

5-Iodo-2-methylpyridine

  • MDL number MFCD07437990
  • Cas Number : 695-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42-51 �C
Synonym 2-Methyl-5-iodopyridine
Purity 97
Storage temp. Store at 0-8
Physical Form White to off-white crystalline solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-iodo-2-methylpyridine/19086?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-iodo-2-methylpyridine/19086?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart