CH6371390052 Chem Impex International, Inc.

2-Chloro-4-hydroxy-3-nitropyridine

  • MDL number MFCD07778466
  • Cas Number : 629655-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-3-nitro-4-pyridinol
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-chloro-4-hydroxy-3-nitropyridine/19092?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-chloro-4-hydroxy-3-nitropyridine/19092?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart