CH6371390180 Chem Impex International, Inc.

3,5-Diamino-2-chloropyridine

  • MDL number MFCD09757507
  • Cas Number : 5632-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122-132 �C
Purity 98
Storage temp. Store at 0-8
Physical Form Beige to coffee color powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-5-diamino-2-chloropyridine/19233?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-5-diamino-2-chloropyridine/19233?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart