CH6371390189 Chem Impex International, Inc.

5-Amino-2-bromo-3-chloropyridine

  • MDL number MFCD09835243
  • Cas Number : 130284-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-3-chloro-5-aminopyridine
Purity 98
Storage temp. Store at 0-8
Physical Form Khaki to brown solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-amino-2-bromo-3-chloropyridine/19243?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-amino-2-bromo-3-chloropyridine/19243?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart