CH6371390211 Chem Impex International, Inc.

(R)-4-Fluorophenylglycine hydrochloride

  • MDL number MFCD09878794
  • Cas Number : 144744-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-4-Fluorophenylglycine hydrochloride
Purity 97
Storage temp. Store at 0-8
Physical Form White to yellow solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-4-fluorophenylglycine-hydrochloride/19266?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-4-fluorophenylglycine-hydrochloride/19266?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart