CH6371390226 Chem Impex International, Inc.

2-Amino-5-bromo-3-ethynylpyrazine

  • MDL number MFCD09999142
  • Cas Number : 1209289-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Physical Form Light brown to yellow crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-amino-5-bromo-3-ethynylpyrazine/19283?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-amino-5-bromo-3-ethynylpyrazine/19283?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart